missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Filmtracer™ LIVE/DEAD™ Biofilm Viability Kit

Product Code. 10480555
Click to view available options
Quantity:
1 kit
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10480555

Brand: Invitrogen™ L10316

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

2 ingredients

Uses mixture of green fluorescent and red fluorescent stains

The FilmTracer LIVE/DEAD Biofilm Viability kit provides a two-color fluorescence assay of bacterial viability, based on membrane integrity, that has proven useful for a diverse array of bacterial genera including those growing in biofilm communities. The LIVE/DEAD Biofilm Viability kit utilizes mixtures of SYTO™ 9 green fluorescent nucleic acid stain and the red-fluorescent nucleic acid stain, propidium iodide. These stains differ both in their spectral characteristics and in their ability to penetrate healthy bacterial cells. When used alone, the SYTO 9 stain generally labels all bacteria in a population: those with intact membranes and those with damaged membranes. In contrast, propidium iodide penetrates only bacteria with damaged membranes, causing a reduction in the SYTO 9 stain fluorescence with both dyes are present. Thus, with an appropriate mixture of the SYTO 9 and propdium iodide stains, bacteria with intact cell membranes stain fluorescent green, whereas bacteria with damaged membranes stain fluorescent red.

Analysis of Bacteria, Bacterial Viability and Cytotoxicity Analysis, Cell Analysis, Microbiological Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Red, Green
Content And Storage 2 ingredients, store in freezer -5°C to -30°C; protect from light
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Confocal Microscope, Fluorescence Microscope, Flow Cytometer, Microplate Reader, Fluorescent Imager
Product Type Biofilm Viability Kit
Dye Type SYTO™ 9, propidium iodide
Emission Visible
Form Liquid
Product Line FilmTracer, Molecular Probes
Quantity 1 kit
Shipping Condition Room Temperature
Sub Cellular Localization Cytoplasm & Cytosol
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.