missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EnzChek™ Reverse Transcriptase Assay Kit

Product Code. 10368542
Change view
Click to view available options
Quantity:
1000 Assays
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10368542 1000 Assays Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10368542 Supplier Invitrogen™ Supplier No. E22064

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

The EnzChek Reverse Transcriptase Assay Kit is a convenient, efficient, and inexpensive assay for measuring reverse transcriptase activity and screening reverse transcriptase inhibitors.

The EnzChek Reverse Transcriptase Assay Kit is a convenient, efficient, and inexpensive assay for measuring reverse transcriptase activity and screening reverse transcriptase inhibitors. In less than an hour, samples can be read in a fluorometer or microplate reader with filter sets appropriate for fluorescein (FITC), GFP, or Alexa Fluor 488.

Features of the EnzChek Reverse Transcriptase Assay Kit include:

  • Detect as little as 0.02 units of HIV-1 reverse transcriptase
  • Large dynamic assay range, detect up to a 50-fold linear range
  • Simple to use assay, amenable for automated high-throughput screening applications

The EnzChek Reverse Transcriptase Assay Kit uses the PicoGreen reagent, which preferentially detects dsDNA or RNA-DNA heteroduplexes over single-stranded nucleic acids or free nucleotides. In this assay, the reverse transcriptase activity in a biological sample generates long RNA-DNA heteroduplexes from a mixture of a long poly(A) template, an oligo-dT primer, and dTTP. The RNA-DNA heteroduplexes formed are then detected by the PicoGreen reagent.

Reverse transcriptase is an enzyme that functions as an RNA-dependent DNA-polymerase to synthesize DNA from an RNA template. Retroviruses such as HIV use reverse transcriptase activity to make a DNA copy of their viral RNA to insert into a host cell genome. Reverse transcriptase enzymes are also used in various cell functions, such as the enzyme telomerase, which maintains telomere length and chromosome stability and has a role in cancer and aging. In molecular biology, reverse transcriptase function is utilized to generate complementary DNA (cDNA) libraries from mRNA and quantify mRNA expression through amplification in RT-PCR.

The EnzChek Reverse Transcriptase Assay Kit provides a simple method to determine reverse transcriptase activity that is useful in the study of retroviruses and screening reverse transcriptase inhibitors, which can function as therapeutic drugs to target retroviral diseases. Because this method is faster and less expensive than standard isotopic assays or immunoassays, it is suitable for testing large numbers of biological samples, enabling automated high-throughput screening of reverse transcriptase inhibitors.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Upon receipt, the kit should be stored at –20°C and protected from light. The 20X TE buffer and the lambda DNA standard are best stored at 4°C; however, either may be frozen for long term storage. Stored properly, the kit's components should remain stable for at least six months. Allow reagents to warm to room temperature before opening the vials.

Detection Method Fluorescence Intensity
For Use With (Application) Reverse Transcriptase Assay
For Use With (Equipment) Fluorescence Microplate Reader
Product Type EnzChek Assay Kit
Product Line EnzChek
Quantity 1000 Assays
Shipping Condition Room Temperature
Sufficient For 1000 Assays
Target Enzyme Reverse Transcriptase

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.