missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EnzChek™ Pyrophosphate Assay Kit

Product Code. 10788224
Change view
Click to view available options
Quantity:
100 Assays
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10788224 100 Assays Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10788224 Supplier Invitrogen™ Supplier No. E6645

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Inorganic pyrophosphatase, 2-Amino-6-mercapto-7-methylpurine riboside (MESG), purine nucleoside phosphorylase (PNP), concentrated reaction buffer, Na2P2O7 standard, and detailed protocols for detecting and quantitating pyrophosphate.

Detects free pyrophosphate in solution

The EnzChek Pyrophosphate Assay Kit provides a rapid and highly sensitive enzymatic assay for detecting free pyrophosphate in solution. In this assay the formation of a chromophoric product is detected spectrophotometrically. The EnzChek Pyrophosphate Detection Kit provides a fast, convenient, and inexpensive spectrophotometric method for measuring the inorganic pyrophosphate produced by a number of biochemical reactions, such as DNA and RNA polymerizations, cyclic AMP formation by the enzyme adenylate cyclase, and the enzymatic activation of fatty acids to form their coenzyme A esters. The EnzChek Pyrophosphate Assay Kit is a modification of our EnzChek Phosphate Assay Kit. In the presence of inorganic phosphate, the substrate 2-amino-6-mercapto-7-methylpurine ribonucleoside (MESG) is converted by the purine nucleoside phosphorylase (PNP) enzyme to ribose 1-phosphate and 2-amino-6-merc apto-7-methylpurine. The enzymatic conversion of MESG results in a shift in absorbance maximum from 330nm to 360nm. The EnzChek Pyrophosphate Assay Kit includes the enzyme inorganic pyrophosphatase, which catalyzes conversion of pyrophosphate into two equivalents of phosphate. The phosphate is then consumed by the MESG/PNP reaction and detected by an increase in absorbance at 360nm. Additional sensitivity is gained by the amplification of one molecule of pyrophosphate into two molecules of phosphate.

  • Sensitivity limit of 1 mM pyrophosphate (approx. 0.2μg/mL)
  • Acommodates a pH range of 6.5 to 8.5

Anion Detection, Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Cellular Imaging, Drug Discovery and Development, Enzyme and Protein Activity Assays, High-Content Screening (HCS), Ionic Homeostasis and Signaling, Phosphatase Activity, Phosphatase Activity Assays, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Ultraviolet
Content And Storage

Includes inorganic pyrophosphatase, 2-Amino-6-mercapto-7-methylpurine riboside (MESG), purine nucleoside phosphorylase (PNP), concentrated reaction buffer, Na2P2O7 standard, and detailed protocols for detecting and quantitating pyrophosphate.

Detection Method Colorimetric
For Use With (Application) Pyrophosphate Assay
For Use With (Equipment) Spectrophotometer
Product Type EnzChek Assay Kit
Dye Type MESG
Format Tube(s)
Product Line EnzChek
Quantity 100 Assays
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.