missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EnzChek™ Phospholipase A2 Assay Kit

Product Code. 10053873
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10053873 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10053873 Supplier Invitrogen™ Supplier No. E10217

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

1 kit

Lower variations due to instrumentation and assay conditions

This kits takes the stand alone assay for phospholipase A2, PLA2 and combines the necessary Ez and lipids to run two complete 96-well microplate assays that monitor activity in purified enzyme preparations and cell lysates. The EnzChek Phospholipase A2 Kit provides enough reagents for two microplates, using 200μL volumes in 96-well format to perform continuous fluorometric monitoring of PLA2 . This product offers an alternative to (B7701), (bis-BODIPY FL C11-PC) reagent, by providing an PLA2 selective substrate and one that is ratiometric, thereby lowering variations produced by instrumentation and assay conditions. Phospholipase A2 or PLA2 represents a family of enzymes that hydrolyze the sn-2 ester linkage of phospholipids. The activities of these enzymes play important roles in cardiovascular, inflammatory and nervous system disorders, and in cancers. The EnzChek Phospholipase A2 substrate provides sensitive and continuous rapid real-time monitoring of PLA2 enzyme activities. This unique substrate is selective for PLA2 and can be used either in a intensity or ratiometric based detection mode. In intensity based detection mode the PLA2 activity is monitored by the intensity increase of a single wavelength at approximately 515nm. In ratiometric analysis, which is based on the distinct fluorescence resonance energy transfer (FRET) emission of this substrate prior to and after cleavage, PLA2 is detected by changes in the emission intensity ratio at 515/575nm with excitation at ≈ 460nm. Either detection mode provides a simple method with low background, high sensitivity and high specificity for PLA2.

Enzyme and Protein Activity Assays, Lipase and Phopholipase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Mixture, Green
Content And Storage 1 kit. Store kit at -<20°, protected from light, desiccated
Detection Method Fluorescence Intensity
For Use With (Application) Phospholipase Assay
For Use With (Equipment) Microplate Reader
Product Type EnzChek Assay Kit
Dye Type BODIPY™ FL
Emission Ratiometric: 460 to 490/515, 575, Intensity-based: 460 to 490/515
Filter Type FITC (Fluorescein) Filter
Product Line EnzChek
Quantity 1 Kit
Shipping Condition Room Temperature
Substrate Properties Lipid-Based Substrate, Chemical Substrate
Substrate Type Phospholipase Substrate
Target Enzyme Phospholipase
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.