missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EnzChek™ Phospholipase A1 Assay Kit

Product Code. 10627215
Change view
Click to view available options
Quantity:
200 Reactions kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10627215 200 Reactions kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10627215 Supplier Invitrogen™ Supplier No. E10219

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

1 kit; Sufficient material is supplied for 200 reactions in 96-well microplates at 100 μL per well as described in the protocol below or 400 reaction in 384 low volume microplates at a volume of ≤25 μL per well.

For continuous monitoring of phospholipase A1 (PLA1) in purified enzyme preparations, cell lysates and living cells

This kit takes the stand-alone assay for phospholipase A1, PLA1 and combines the necessary reagents to run 2 to 10 complete 96-well microplate assays that monitor activity in purified enzyme preparations and cell lysates. The importance of phospholipases in cellular signaling, lipid metabolism, inflammatory responses and pathological disorders related to these processes has stimulated demand for fluorescence-based activity monitoring methods. In particular the phospholipases resident in plasma and endothelium can perturb circulating LDL and HDL particles, creating pro-artherogenic forms. Recent evidence is drawing a link between these lipases and the progression of several severe neurodegenerative diseases, including Alzheimer's. PLA-1 improves upon two existing reagents in two ways: first it is now PLA-1 specific and secondly it has improved the assay quality by decreasing initial background noise.

Enzyme and Protein Activity Assays, Lipase and Phopholipase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 kit. Store kit at -<20°C, protected from light, desiccated

Sufficient material is supplied for 200 reactions in 96 well microplates at 100 μl per well as described in the protocol below or 400 reaction in 384 low volume microplates at a volume of ≤ 25 μL per well.
Detection Method Fluorescence Intensity
For Use With (Application) Phospholipase Assay
Product Type EnzChek Assay Kit
Product Line EnzChek
Quantity 200 Reactions kit
Shipping Condition Room Temperature
Substrate Properties Lipid-Based Substrate, Chemical Substrate
Substrate Type Phospholipase Substrate
Target Enzyme Phospholipase

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.