missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EnzChek™ Phosphate Assay Kit

Product Code. 10686333
Change view
Click to view available options
Quantity:
100 Assays
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10686333 100 Assays Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10686333 Supplier Invitrogen™ Supplier No. E6646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Determine phosphate released during ATPase or GTPase enzymatic reactions

The EnzChek Phosphate Assay Kit provides a fast and sensitive spectrophotometric method for the quantification of inorganic phosphate in solution and for phosphate released during ATPase or GTPase enzymatic reactions. In the presence of inorganic phosphate, the 2-amino-6-mercapto-7-methylpurine riboside (MESG) substrate is converted by the purine nucleoside phosphorylase (PNP) enzyme to the ribose 1-phosphate and 2-amino-6-mercapto-7-methylpurine product. This enzymatic conversion of MESG results in a spectrophotometric shift in maximum absorbance from 330nm for the substrate to 360nm for the product. The reaction can be performed over a pH range of 6.5 to 8.5. The EnzChek phosphate reaction is sufficiently fast and quantitative for use in stopped-flow kinetic experiments.

  • Sensitivity of 2 to 150μM phosphate
  • Spectrophotometric microplate-based assay detects a change in absorbance maximum from 330nm to 360nm
  • Continuous monitoring of phosphate generated by ATPase or GTPase activity

Anion Detection, Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Cellular Imaging, Drug Discovery and Development, Enzyme and Protein Activity Assays, High-Content Screening (HCS), Ionic Homeostasis and Signaling, Phosphatase Activity, Phosphatase Activity Assays, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Ultraviolet
Content And Storage Store in freezer -5°C to -30°C.
Detection Method Colorimetric
For Use With (Application) Phosphate Assay
For Use With (Equipment) Spectrophotometer
Product Type EnzChek Assay Kit
Dye Type MESG
Format Tube(s)
Product Line EnzChek
Quantity 100 Assays
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.