missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DiIC16(3) (1,1'-Dihexadecyl-3,3,3',3'-Tetramethylindocarbocyanine Perchlorate)

Product Code. 11570316
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11570316 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11570316 Supplier Invitrogen™ Supplier No. D384

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

When applied to cells, the dye diffuses laterally within the plasma membrane

The red-orange fluorescent, lipophilic carbocyanine DiIC16(3) is a shorter-chain DiI (DiIC18(3)) analog that may incorporate into membranes more easily than DiI. It is weakly fluorescent in water but highly fluorescent and quite photostable when incorporated into membranes. It has an extremely high extinction coefficient and short excited-state lifetimes (∽1 nanosecond) in lipid environments.

Cell Analysis, Cell Tracing & Tracking, Neuronal Tracing

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Content And Storage Store at room temperature and protect from light.
For Use With (Equipment) Fluorescence Microscope
Quantity 100 mg
Detection Method Fluorescence
Shipping Condition Room Temperature
Product Type Liphophilic Tracer
Sub Cellular Localization Cell Membranes, Lipids

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.