missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dextran, Oregon Green™ 488; 10,000 MW, Anionic, Lysine Fixable

Product Code. 11534716
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11534716

Brand: Invitrogen™ D7171

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Rigorous methods for removing as much unconjugated dye as practical, followed by assay of dextran conjugates by thin-layer chromatography to help ensure absence of low molecular weight contaminants

Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of live cells, and to report the hydrodynamic properties of the cytoplasmic matrix. The labeled dextran is commonly introduced into the cells via microinjection. Dextrans are hydrophilic polysaccharides characterized by their moderate-to-high molecular weight, good water solubility, and low toxicity. They also generally exhibit low immunogeniticy. Dextrans are biologically inert due to their uncommon poly-(α-D-1,6-glucose) linkages, which render them resistant to cleavage by most endogenous cellular glycosidases.

  • Label (Ex/Em): Oregon Green™ 488 (496/524)
  • Size: 10,000 MW
  • Charge: Anionic
  • Fixable: Fixable via Lysine
  • Succinimidyl coupling of dyes to dextran molecule, which, in most cases, results in neutral or anionic dextran
  • Reaction used to produce the Rhodamine Green™ and Alexa Fluor™ 488 dextrans results in the final product being neutral, anionic, or cationic
  • Common uses include neuronal tracing (anterograde and retrograde) in live cells
  • Cell lineage tracing in live cells
  • Neuroanatomical tracing
  • Examines intercellular communications (e.g., in gap junctions, during wound healing, and during embryonic development)
  • Investigates vascular permeability and blood-brain barrier integrity
  • Tracks endocytosis
  • Monitors acidification (some dextran-dye conjugates are pH-sensitive)
  • Studies hydrodynamic properties of cytoplasmic matrix

  • Oregon Green dextrans are intrinsically anionic
  • To produce more highly anionic dextrans, proprietary procedure adds negatively charged groups to dextran carriers; these products are designated polyanionic dextrans
  • Some applications require that dextran tracer be treated with formaldehyde or glutaraldehyde for subsequent analysis: For these applications, lysine-fixable dextran conjugates of fluorophores or biotin
  • Dextrans have covalently bound lysine residues that permit dextran tracers to be conjugated to surrounding biomolecules by aldehyde-mediated fixation for subsequent detection by immunohistochemical and ultrastructural techniques

Cell Analysis, Cell Tracing and Tracking, General Cell Tracing, Neuronal Tracing

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type Dextran
Quantity 5 mg
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label or Dye Other Label(s) or Dye(s)
Shipping Condition Room Temperature
Product Line Oregon Green

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.