Learn More
Invitrogen™ Dextran, Amino, 10,000 MW
Description
Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of live cells, and to report the hydrodynamic properties of the cytoplasmic matrix. The labeled dextran is commonly introduced into the cells via microinjection. Dextrans are hydrophilic polysaccharides characterized by their moderate-to-high molecular weight, good water solubility, and low toxicity. They also generally exhibit low immunogeniticy. Dextrans are biologically inert due to their uncommon poly-(α-D-1,6-glucose) linkages, which render them resistant to cleavage by most endogenous cellular glycosidases.
- Succinimidyl coupling of dyes to dextran molecule, which, in most cases, results in neutral or anionic dextran
- Suitable for neuronal tracing (anterograde and retrograde) in live cells
- Cell lineage tracing in live cells
- Examining intercellular communications (e.g., in gap junctions, during wound healing, and during embryonic development)
- Investigating vascular permeability and blood-brain barrier integrity
- Monitoring acidification (some dextran-dye conjugates are pH-sensitive)
- Studying hydrodynamic properties of cytoplasmic matrix
Cell Analysis, Cell Tracing and Tracking, General Cell Tracing, Neuronal Tracing
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Product Type | Dextran |
| Quantity | 1 g |
| Content And Storage | Store at room temperature. |
| Label or Dye | Unlabeled |
| Shipping Condition | Room Temperature |
| Product Line | Invitrogen |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.