missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DAF-FM Diacetate (4-Amino-5-Methylamino-2',7'-Difluorofluorescein Diacetate)

Product Code. 10082632
Click to view available options
Quantity:
1 mg
10 x 50 μg
Unit Size:
1mg
500µg
This item is not returnable. View return policy

Product Code. 10082632

Brand: Invitrogen™ D23842

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For detection and quantification of low concentrations of nitric oxide (NO)

It is essentially nonfluorescent until it reacts with NO to form a fluorescent benzotriazole. DAF-FM fluorescence can be detected by any instrument that can detect fluorescein, including flow cytometers, microscopes, fluorescent microplate readers and fluorometers.

  • Ex/Em of DAF-FM: approx. 495/515nm
  • Lyophilized product should be dissolved using DMSO and then added to an aqueous buffer to create a working solution
  • DAF-FM diacetate is cell permeant and passively diffuses across cellular membranes; once inside the cell, it is converted to cell-impermeant for
  • Buffers containing bovine serum albumin (BSA) and phenol red may affect fluorescence and should be used with caution
  • Fluorescence quantum yield of DAF-FM is approx. 0.005, but increases about 160-fold, to ∼0.81, after reacting with NO
  • Assessment of NO production in transaldolase-deficient lymphoblasts by flow cytometry
  • Detection of NO accumulation in embryonic cortical neurons following neurotrophin stimulation
  • in vivo imaging of NO in zebrafish
I

  • ntravital microscopic detection of NO generation associated with angiogenesis in mice
  • Quantitation of ATP-induced NO release in rabbit platelets
  • Spectra of the NO adduct of DAF-FM are independent of pH above pH 5.5
  • NO adduct of DAF-FM is significantly more photostable than that of DAF-2, which means additional time for image capture
  • DAF-FM is a more sensitive reagent for NO than is DAF-2 (NO detection limit for DAF-FM approx. 3nM versus approx. 5nM for DAF-2)

Refer to the Molecular Probes™ Handbook for additional product information.

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Nitric Oxide Research, Nitro-Oxidative Stress

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Product Type NO Indicator

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.