missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Click-IT™ L-Homopropargylglycine (HPG)

Product Code. 10328924
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10328924 5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10328924 Supplier Invitrogen™ Supplier No. C10186
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Less hazardous technique for the detection of nascent protein synthesis

Click-iT homopropargylglycine (HPG) provides a fast, sensitive, non-toxic and most importantly non-radioactive alternative to the traditional technique, 35S methionine, for the detection of nascent protein synthesis. HPG is an amino acid analog that contains a very small modification, specifically an alkyne moiety that can be fed to cultured cells and incorporated into proteins during active protein synthesis. Detection utilizes the chemoselective ligation or “click” reaction between and azide and an alkyne where the alkyne modified protein is detected with an azide-containing dye or hapten together with either the Click-iT Cell Reaction Buffer Kit or the Click-iT Protein Buffer Kit. With the Click-iT Cell Reaction Buffer Kit, cells can be analyzed by fluorescence microscopy, flow cytometry or high-content imaging and analysis. With the Click-iT Protein Reaction Buffer Kit, achieve detection sensitivity in 1-D gels and western blots in the low femtomole range or perform LC-MS/MS and MALDI MS analysis.

  • Less hazardous

Cell Analysis, Click Chemistries, Glycoprotein Analysis with Click Chemistry, Labeling Chemistry, Metabolic Labeling and Nascent Protein Synthesis with Click Chemistry

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type L-Homopropargylglycine
Quantity 5 mg
Content And Storage Store at ≤-20°C and desiccated.
Labeling Method Metabolic Labeling
Shipping Condition Room Temperature
Product Line Click-iT, Molecular Probes
Label or Dye Alkyne
Format Solid
Labeling Target Proteins

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.