missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Click-IT™ GlcNAz Metabolic Glycoprotein Labeling Reagent (tetraacetylated N-Azidoacetylglucosamine)
GREENER_CHOICE

Product Code. 10716783
Change view
Click to view available options
Quantity:
5.2 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10716783 5.2 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10716783 Supplier Invitrogen™ Supplier No. C33367

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Compatible with LC-MS/MS and Multiplexed Proteomics™ technologies

The Click-iT GlcNAz metabolic glycoprotein labeling reagent provides the first part of a simple and robust two-step technique to identify and characterize intracellular O-GlcNAc glycoproteins. In step one, cultured cells are incubated with the azide-modified glucosamine (GlcNAc). The azido-sugar is incorporated into intracellular O-GlcNAc-containing glycoproteins through the permissive nature of the oligosaccharide biosynthesis pathway. In step two, via the chemoselective ligation or click reaction between an azide and an alkyne, the azido-labeled glycoproteins can then detected with a Click-iT Glycoprotein Detection kit for gels (TAMRA or Dapoxyl™ alkyne) or Western blots (biotin alkyne). The Click-iT glycoprotein products are compatible with LC-MS/MS and Multiplexed Proteomics™ technologies for in-depth analyses of the glycoproteome.

  • Less hazardous

Cell Analysis, Click Chemistries, Glycoprotein Analysis with Click Chemistry, Labeling Chemistry, Mass Spectrometry, Metabolic Labeling, Peptide Labeling, Protein and Antibody Labeling, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type Labeling Reagent
Quantity 5.2 mg
Content And Storage Store in freezer -5°C to -30°C.
Detection Method Biotin-based, Fluorescence
Shipping Condition Room Temperature
Product Line Click-iT
Label or Dye Alexa Fluor 488, Alexa Fluor 555, Alexa Fluor 594, Alexa Fluor 647, Biotin, Oregon Green 488, TMR (Tetramethylrhodamine)
Labeling Target Proteins

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.