Learn More
Invitrogen™ Click-IT™ GalNAz Metabolic Glycoprotein Labeling Reagent (Tetraacetylated N-Azidoacetylgalactosamine)

Description
The Click-iT GalNAz metabolic glycoprotein labeling reagent provides the first part of a simple and robust two-step technique to identify and characterize cell surface O-linked glycoproteins. In step one, cultured cells are incubated with the azide-modified galactosamine (GalNAz). The azido-sugar is metabolically incorporated into cell surface O-linked glycoproteins through the permissive nature of the oligosaccharide biosynthesis pathway. In step two, via the chemoselective ligation or click reaction between an azide and an alkyne, the azido-labeled glycoproteins can then be detected with a Click-iT Glycoprotein Detection Kit for gels (TAMRA or Dapoxyl™ alkyne) or Western blots (biotin alkyne). These Click-iT products are compatible with LC-MS/MS and Multiplexed Proteomics™ technologies for in-depth analyses of the glycoproteome.
- Less hazardous
Cell Analysis, Click Chemistries, Glycoprotein Analysis with Click Chemistry, Labeling Chemistry, Mass Spectrometry, Metabolic Labeling, Peptide Labeling, Protein and Antibody Labeling, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Product Type | Labeling Reagent |
| Quantity | 5.2 mg |
| Content And Storage | Store in freezer -5°C to -30°C. |
| Detection Method | Biotin-based, Fluorescence |
| Shipping Condition | Room Temperature |
| Product Line | Click-iT |
| Label or Dye | Alexa Fluor 488, Alexa Fluor 555, Alexa Fluor 594, Alexa Fluor 647, Biotin, Oregon Green 488, TMR (Tetramethylrhodamine) |
| Labeling Target | Proteins |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.