missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Molecular Probes™ Click-iT™ EdU Microplate Assay

Product Code. 10286453
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10286453 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10286453 Supplier Invitrogen™ Supplier No. C10214

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Kit contains sufficient reagents for performing 400 individual assays in a 96-well plate format.

Simple and rapid workflow

The Click-iT EdU Microplate Assay is a superior alternative to traditional cell proliferation assays that is optimized for microplate-based fluorescence analysis. This assay provides a simple and rapid workflow with reduced wash steps compared to traditional BrdU colorimetric or fluorescent proliferation assays. In this assay the modified thymidine analogue EdU is efficiently incorporated into newly synthesized DNA and fluorescently labeled with bright, photostable Amplex UltraRed dye in a fast, highly specific click reaction. This fluorescent labeling of proliferating cells is accurate and compatible with antibody methods due to the mild click protocol. The most accurate proliferation-detection methods are based on the incorporation and measurement of nucleoside analogues in newly synthesized DNA, with bromodeoxyuridine (BrdU) a commonly used analogue. BrdU-labeled DNA is quantitated using anti-BrdU antibodies following DNA denaturation by harsh methods (HCl, heat, or enzymes) to expose the BrdU molecules. This step is time consuming and difficult to perform consistently.

  • Simple: works the first time, every time, in less time
  • Efficient: no denaturation steps or harsh treatment required
  • Sensitive: outstanding sensitivity and dynamic range
  • Versatile: fluorescence or absorbance readouts through use of Amplex UltraRed reagent
  • Less hazardous

Superior Proliferation Methodology
The Click-iT EdU Microplate Assay provides a superior alternative to BrdU assays for measuring cell proliferation. EdU (5-ethynyl-2'-deoxyuridine) is a nucleoside analog of thymidine and is incorporated into DNA during active DNA synthesis. With Click-iT EdU, mild fixation and detergent permeabilization is sufficient for the small molecule-based Click-iT EdU detection reagent to gain access to the DNA. The signal amplification steps include incubation with an anti-Oregon Green™ antibody conjugated to horseradish peroxidase (HRP), which then reacts with Amplex UltraRed substrate in a 1:1 stoichiometry and produces a bright red, fluorescent product (excitation/emission maxima approx. 568/585 nm). With a high extinction coefficient, good quantum efficiency, and resistance to auto-oxidation, Amplex UltraRed reagent delivers higher sensitivity and a broader assay range than other fluorogenic or colorimetric peroxidase substrates. Amplex UltraRed reagent also offers versatility through detection by either fluorescence or absorbance measurement. This kit also includes Amplex UltraRed stop reagent that enables the peroxidase reaction to be terminated at a user-determined time point and stabilizes the signal for up to 24 hours. As a consequence, the Click-iT EdU Microplate Assay is not only easy to use, but more accurate and compatible with cell cycle analysis and other intracellular or extracellular targets for truly content-rich results.

Cell Analysis, Cell Proliferation, Cell Proliferation with Click Chemistry, Cell Viability, Proliferation and Function, Click Chemistries, Labeling Chemistry

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 kit
Detection Method Fluorescence
Product Type Microplate Assay
Conjugate Amplex UltraRed
Kit Contents Ssufficient reagents for performing 400- individual assays in a 96-well plate format
Format 96-well Plate
Color Red
Content And Storage Store kit at -20°C, desiccated and protected from light. When stored as directed, this kit is stable for 1 year.
Excitation/Emission 568/585
For Use With (Equipment) Microplate Reader

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.