missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Click-IT™ Biotin Protein Analysis Detection Kit
GREENER_CHOICE

Product Code. 10597733
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10597733 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10597733 Supplier Invitrogen™ Supplier No. C33372

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Identify and characterize glycoproteins by Western blot

The Click-iT Biotin Glycoprotein Detection Kit provides the second part of a simple and robust technique to identify and characterize glycoproteins by Western blot. In step two, after the incorporation of an azide moiety into protein glycan structures with either a Click-iT metabolic labeling reagent or the Click-iT Enzymatic Labeling system, the azide-modified glycoproteins are detected via the chemoselective ligation or click reaction between an azide and an alkyne. With this technique, detection sensitivity is in the low femptomole range and biotin-labeled samples can be detected before or after probing the Western with a primary antibody.

  • Less hazardous

Cell Analysis, Click Chemistries, Glycoprotein Analysis with Click Chemistry, Labeling Chemistry, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis, Western Blotting

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type Biotin Protein Analysis Detection Kit
Quantity 1 Kit
Content And Storage Store in freezer -5°C to -30°C.
Detection Method Biotin-based, Fluorescence
Shipping Condition Room Temperature
Product Line Click-iT
Label or Dye Biotin
Labeling Target Proteins

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.