missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Click-iT™ AHA Alexa Fluor™ 488 Protein Synthesis HCS Assay
GREENER_CHOICE

Product Code. 10074833 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 plates
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10074833 2 plates Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10074833 Supplier Invitrogen™ Supplier No. C10289
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

The kit contains sufficient reagents for performing two plates in a 96-well plate format.

Non-radioactive technique

The Click-iT AHA Alexa Fluor 488 Protein Synthesis HCS Assay provides a fast, sensitive, non-toxic and non-radioactive method for the detection of nascent protein synthesis utilizing fluorescence microscopy and high-content imaging. Click-iT AHA is an amino acid analog of methionine containing an azido moiety. Similar to 35S-methionine, Click-iT AHA is fed to cultured cells and incorporated into proteins during active protein synthesis. Detection of the incorporated amine acid utilizes a chemoselectiove ligation or 'click' reaction between an azide and an alkyne, where the azido modified protein is detected with the green-fluorescent Alexa Fluor 488 alkyne. The Click-iT AHA Alexa Fluor 488 Protein Synthesis HCS Assay has been successfully tested in A549 and U-2 OS cells with a variety of reagents that inhibit protein synthesis including cycloheximide, anisomycin and puromycin in both dose response and Min/Max format.

  • Less hazardous

Cell Analysis, Cell Metabolism, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Blue, Green
Content And Storage The kit contains sufficient reagents for performing 2 plates in a 96-well plate format.
  • Store at ≤-20°C, desiccate and protect from light.
  • Detection Method Fluorescence
    For Use With (Application) High-content Screening (HCS)
    For Use With (Equipment) Fluorescence Microscope, High Content Analysis Instrument
    Product Type HCS Assay
    Dye Type Alexa Fluor™ 488, Hoechst 33258
    Format 96-well plate
    Product Line Alexa Fluor, Click-iT, Molecular Probes
    Quantity 2 plates
    Shipping Condition Room Temperature
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.