missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellLight™ Lysosomes-GFP, BacMam 2.0

Product Code. 10304759
Click to view available options
Quantity:
100μL
1 mL
Unit Size:
100µL
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10304759

Brand: Invitrogen™ C10596

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Provides easy method for labeling of lysosomes with Green Fluorescent Protein in live cells

This ready-to-use construct is transduced/transfected into cells using BacMam 2.0 technology, where it expresses GFP fused to Lamp1 (lysosomal associated membrane protein 1). You can observe lysosomes-GFP behavior in live cells using a fluorescent imaging system and standard FITC/GFP filter set. Study staining of lysosomes, which does not depend upon organelle pH or some other parameter, that can be different from one cell type to other. In addition, you can co-transduce/transfect more than one CellLight reagent and label live cells with multiple tracking or tracing dyes to image dynamic cellular processes. Cells expressing CellLight constructs can also be fixed with formaldehyde for multiplexing with antibodies using immunocytochemical techniques.

BacMam Technology

CellLight Lysosomes-GFP, BacMam 2.0, is a fusion construct of Lamp1 (lysosomal associated membrane protein 1) and emGFP, providing accurate and specific targeting to cellular lysosomes-GFP. This fusion construct is packaged in the insect virus baculovirus, which does not replicate in human cells and is designated as safe to use with biosafety level (BSL) 1 in most laboratories. BacMam technology ensures that most mammalian cell types are transduced/transfected with high efficiency and minimal toxicity. This transient transfection can be detected after overnight incubation for up to five days—enough time to carry out most dynamic cellular analyses. Like any transfection/transduction technique, the BacMam method does not transfect/transduce all of the cells with equal efficiency, making it poorly suited to cellular population studies or automated imaging/counting. CellLight reagents are ideal for experiments where cellular or subcellular co-locatization is required, or for cellular function studies that need special resolution.

  • Highly efficient: up to 90% transduction of wide range of mammalian cell lines, including primary cells, stem cells, and neurons
  • Fast and convenient: simply add CellLight reagent to cells, incubate overnight, and image—or store frozen, assay-ready cells for later use
  • Less toxic: CellLight reagents are non-replicating in mammalian cells and are suitable for biosafety level (BSL) 1 handling
  • Flexible: co-transduce more than one BacMam reagent for multiplex experiments or co-localization studies; tightly control expression levels by simply varying the dose
  • Flow Cytometer Laser Lines: 488

Cell Analysis, Cell Structure, Cell Viability, Proliferation & Function, Cellular Imaging, Chemotaxis & Cell Migration, Cytoskeleton, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Staining & Detection

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Content And Storage Store at 2°C to 6°C, protected from light. Do Not Freeze.
Excitation Wavelength Range 488⁄510
Dye Type GFP (EmGFP)
For Use With (Equipment) Confocal Microscope, Fluorescence Microscope
Quantity 1 mL
Detection Method Fluorescence
Emission Visible
Form Liquid
Product Line CellLight
Shipping Condition Wet Ice
Technique Fluorescence Intensity
Label Type Fluorescent Dye
Product Type Lysosomal Probe
SubCellular Localization Lysosomes
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.