missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellLight™ Late Endosomes-GFP, BacMam 2.0

Product Code. 12313613
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12313613 1 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12313613 Supplier Invitrogen™ Supplier No. C10588

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provides easy way to label late endosomes with green fluorescent protein in live cells

This ready-to-use construct is transfected into cells using BacMam 2.0 technology, where it expresses GFP fused to Rab7a. You can observe late endosomes-GFP behavior in live cells independently of organelle pH and label with multiple tracking or tracing dyes to image dynamic cellular processes.

Cells expressing CellLight constructs can also be fixed with formaldehyde for multiplexed imaging using immunocytochemical techniques.

BacMam Technology

CellLight Late Endosomes-GFP, BacMam 2.0, is a fusion construct of Rab7a and emGFP, providing accurate and specific targeting to cellular Late Endosomes-GFP. This fusion construct is packaged in the insect virus baculovirus, which does not replicate in human cells and is designated as safe to use with biosafety level (BSL) 1 in most laboratories. BacMam technology ensures that most mammalian cell types are transduced/transfected with high efficiency and minimal toxicity. This transient transfection can be detected after overnight incubation for up to five days—enough time to carry out most dynamic cellular analyses. Like any transfection/transduction technique, the BacMam method does not transfect/transduce all of the cells with equal efficiency, making it poorly suited to cellular population studies or automated imaging/counting. CellLight reagents are ideal for experiments where cellular or subcellular co-locatization is required, or for cellular function studies that need special resolution.

  • Fast and convenient: simply add CellLight reagent to cells, incubate overnight, and image—or store frozen, assay-ready cells for later use
  • Highly efficient: up to 90% transduction of a wide range of mammalian cell lines, including primary cells, stem cells, and neurons
  • Flexible: co-transduce more than one BacMam reagent for multiplex experiments or co-localization studies; tightly control expression levels by simply varying the dose
  • Less toxic: CellLight reagents are non-replicating in mammalian cells and are suitable for biosafety level (BSL) 1 handling

Cell Analysis, Cell Structure, Cell-Based Reporter Assays, Cellular Imaging, Endosomes, Fluorescent Protein Assays (GFP), High-Content Screening (HCS), Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Staining & Detection, In Vivo Imaging

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Content And Storage 2°C to 6°C, protect from light. Do not freeze.
Excitation Wavelength Range 488⁄510
Dye Type GFP (EmGFP)
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer, Fluorescent Imager
Quantity 1 mL
Detection Method Fluorescence
Emission Visible
Form Liquid
Product Line CellLight, Molecular Probes
Shipping Condition Wet Ice
Target Function Endocytosis
Label Type Fluorescent Dye
Product Type Endosomes Label
SubCellular Localization Endosomes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.