missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Carboxy-H2DFFDA

Product Code. 11524227
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11524227 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11524227 Supplier Invitrogen™ Supplier No. C13293
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Chemically reduced, acetylated form of fluorescein indicatesr reactive oxygen species in cells.

H2DFFDA is nonfluorescent molecule that readily converts to green-fluorescent form when acetate groups are removed by intracellular esterases and oxidation (by the activity of ROS) occurs within the cell. This fluorinated analog exhibits improved photostability compared to the chlorinated fluorescein derivatives (DCF, DCFDA).

  • Ex/Em: approx. 492–495/517–527nm
  • Product is air sensitive (stored under dry argon or nitrogen)
  • Product may be dissolved in DMSO, DMF, or ethanol for use
  • Indicator is cell permeant
  • Fluorescence can be monitored using a flow cytometer, fluorometer, microplate reader, or fluorescence microscope, using excitation sources and filters appropriate for fluorescein

Refer to the Molecular Probes™ Handbook for additional product information.

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Nitro-Oxidative Stress, Reactive Oxygen Species

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Detection Method Fluorescence
Product Type ROS Indicator

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.