missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calcium Green™-1, AM, cell permeant
Description
- They have uses in many calcium signaling investigations, including measuring intracellular Ca2+, following Ca2+ influx and release, and multiphoton excitation imaging of Ca2+ in living tissues
- Cells may be loaded with the AM ester forms of these calcium indicators by adding dissolved indicator directly to dishes containing cultured cells
- Fluorescence signal from these cells is generally measured using fluorescence microscopy, fluorescence microplate assays, or flow cytometry
- Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength
- These probes are excited by visible light, and because energy required for excitation is low, potential for cellular photodamage is reduced
- Commonly used laser-based instruments (i.e., confocal laser scanning microscopes) are able to efficiently excite these indicators, and their emissions are in regions of spectrum where cellular autofluorescence and scattering backgrounds are often less of a problem
- Label (Ex/Em): Calcium Green; -1 (506/531nm)
- Fluorescence intensity increase upon binding Ca2+ : approximately 14 fold
Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Application) | Cell Viability and Proliferation |
| For Use With (Equipment) | Fluorescence Microscope |
| Product Type | Calcium Indicator |
| Dye Type | Fluorescent Dye-Based |
| Product Line | Calcium Green |
| Quantity | 500 μg |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?