missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Calcium Green™-1, AM, cell permeant

Product Code. 11588936
Change view
Click to view available options
Quantity:
10 x 50 μg
500 μg
Unit Size:
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11588936 10 x 50 μg 500µg
11554257 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11588936 Supplier Invitrogen™ Supplier No. C3012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+

  • They have uses in many calcium signaling investigations, including measuring intracellular Ca2+, following Ca2+ influx and release, and multiphoton excitation imaging of Ca2+ in living tissues
  • Cells may be loaded with the AM ester forms of these calcium indicators by adding dissolved indicator directly to dishes containing cultured cells
  • Fluorescence signal from these cells is generally measured using fluorescence microscopy, fluorescence microplate assays, or flow cytometry
  • Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength
  • These probes are excited by visible light, and because energy required for excitation is low, potential for cellular photodamage is reduced
  • Commonly used laser-based instruments (i.e., confocal laser scanning microscopes) are able to efficiently excite these indicators, and their emissions are in regions of spectrum where cellular autofluorescence and scattering backgrounds are often less of a problem
  • Label (Ex/Em): Calcium Green; -1 (506/531nm)
  • Fluorescence intensity increase upon binding Ca2+ : approximately 14 fold

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Cell Viability and Proliferation
For Use With (Equipment) Fluorescence Microscope
Product Type Calcium Indicator
Dye Type Fluorescent Dye-Based
Product Line Calcium Green
Quantity 10 x 50 μg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.