Learn More
Invitrogen™ Molecular Probes™ BODIPY™ Vinblastine, BODIPY™ FL Conjugate (BODIPY FL Vinblastine)
Fluorescent analog of vinblastine
Brand: Invitrogen™ V12390
Code : 50
Additional Details : Weight : 0.05250kg
Description
BODIPY FL vinblastine, a fluorescent analog of the anticancer drug vinblastine, may be a useful probe for labeling β-tubulin or for investigating drug-transport mechanisms.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining and Detection
Order Info
Shipping Condition: Room temperature
Specifications
Probe, Tubulin Selective | |
100 μg | |
Store in freezer (-5°C to -30°C) and protect from light. |
For Research Use Only. Not for use in diagnostic procedures.