Learn More
Invitrogen™ CellTrace™ BODIPY™ TR Methyl Ester (Lipophilic Counterstain For GFP)
Retained after paraformaldehyde fixation
Brand: Invitrogen™ C34556
324.52 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00525kg
Description
- Readily permeates cell membranes and selectively stains mitochondria and endomembranous organelles such as endoplasmic reticulum and Golgi apparatus, but does not appear to localize in plasma membrane
Cell Adhesion, Cell Analysis, Cell Proliferation, Cell Tracing and Tracking, Cell Viability, Proliferation and Function, Cell-Based Reporter Assays, Cellular Imaging, Cellular Toxicology Assays, Chemotaxis and Cell Migration, Drug Discovery and Development, Enzyme and Protein Activity Assays, Fluorescent Protein (e.g. GFP) Assays, Fluorescent Protein Assays (GFP), General Cell Tracing, High-Content Screening (HCS), Immunofluorescence (IF), Immunofluorescence Staining and Detection, Proteins, Expression, Isolation and Analysis, Reporter Assays, Target-Based ADME/Tox Assays
Order Info
Shipping Condition: Room temperature
Specifications
Store in freezer -5°C to -30°C and protect from light. | |
BODIPY™, CellTrace™ | |
Room Temperature | |
Lipophilic Tracer |
BODIPY Dyes | |
Cell Tracker Compounds, Cell Labeling Reagents | |
Other Labels or Dyes |
For Research Use Only. Not for use in diagnostic procedures.