Learn More
Invitrogen™ BAPTA, Tetrapotassium Salt, cell impermeant
Highly selective chelator for Ca2+ over Mg2+ and can be used to control level of extracellular Ca2+
Brand: Invitrogen™ B1204
190.76 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00525kg
Description
- BAPTA is more selective for Ca2+ than EDTA and EGTA, and its metal binding is also less sensitive to pH
Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling
Order Info
Shipping Condition: Room temperature
Specifications
Store at room temperature. | |
BAPTA Tetrapotassium Salt | |
Chelator (Non-Fluorescent) | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.