missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Amplex™ Red Phosphatidylcholine-Specific Phospholipase C Assay Kit

Product Code. 10023302
Change view
Click to view available options
Quantity:
500 Assays
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10023302 500 Assays Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10023302 Supplier Invitrogen™ Supplier No. A12218
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Detect phosphatidylcholine-specific phospholipase C (PC-PLC) activity using a fluorescence microplate reader or fluorometer

The Amplex Red Phosphatidylcholine-Specific Phospholipase C Assay Kit provides a sensitive method for detecting phosphatidylcholine-specific phospholipase C (PC-PLC) activity using a fluorescence microplate reader or fluorometer. In this enzyme-coupled assay, PC-PLC activity is monitored indirectly using 10-acetyl-3,7-dihydroxyphenoxazine (Amplex Red reagent), a sensitive fluorogenic probe for hydrogen peroxide. PC-PLC converts phosphatidylcholine (lecithin) to phosphocholine and diacylglycerol. Alkaline phosphatase is added, which hydrolyzes phosphorylcholine to form choline. Choline is then oxidized by choline oxidase to form betaine and hydrogen peroxide. In the presence of horseradish peroxidase, the hydrogen peroxide reacts with Amplex Red reagent in a 1:1 stoichiometric ratio to generate the highly fluorescent product resorufin. Because resorufin has absorption and fluorescence emission maxima of approximately 571nm and 585nm, respectively, there is little interference from autofluorescence in most biological samples.

  • Detects phospholipase-C activity levels as low as 0.2mU/mL
  • Format allows for multiple time point measurements
  • Designed for minimal autofluorescence interference
  • Custom assay design and packaging are also available

Use Amplex Red Assays for a Broad Range of Investigations

A wide variety of validated Amplex Red assays are available for studying cell signaling and lipids, neurobiology, inflammation and immune function, and metabolism. We also offer Amplex UltraRed Reagent, a second-generation reagent providing greater sensitivity and brighter fluorescence, and the Amplex Red/UltraRed Stop Reagent. The Amplex Red/UltraRed Stop Reagent provides convenience and control by allowing the fluorescence signal-generating reaction to be terminated at a user-determined time point. After addition of the stop reagent, the fluorescence signal remains stable for at least three hours.

Enzyme and Protein Activity Assays, Lipase and Phopholipase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Phospholipase Assay
For Use With (Equipment) Fluorometer, Microplate Reader
Product Type Amplex Red Assay Kit
Dye Type Other Label(s) or Dye(s)
Format Cuvettes, 96-well plate
Product Line Amplex
Quantity 500 Assays
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.