missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Amplex™ Red Neuraminidase (Sialidase) Assay Kit

Product Code. 10594993
Click to view available options
Quantity:
400 Assays
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10594993

Brand: Invitrogen™ A22178

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Format allows for multiple time point measurements

The Amplex Red Neuraminidase (Sialidase) Assay Kit provides a sensitive and simple method for detecting neuraminidase activity in serum and in purified enzyme systems using either a fluorescence microplate reader or fluorometer. Neuraminidase (also known as sialidase) is a very common enzyme that hydrolyzes terminal sialic acid residues on polysaccharide chains, most often exposing a galactose residue. Neuraminidase plays a key role in the invasion of target cells and the subsequent replication of the influenza virus, making neuraminidase an important target for influenza drug development. The Amplex Red Neuraminidase (Sialidase) Assay Kit provides an ultrasensitive method for detecting neuramimidase activity. This assay utilizes Amplex Red reagent to detect hydrogen peroxide generated by galactose oxidase oxidation of desialiated galactose, the end result of neuraminidase action. Then the hydrogen peroxide, in the presence of horseradish peroxidase (HRP), reacts in a 1:1 stoichiometric ratio with Amplex Red reagent to generate the red-fluorescent oxidation product resorufin. Because resorufin has absorption and fluorescence emission maxima of approximately 571nm and 585nm, respectively, there is little interference from autofluorescence in most biological samples.

  • Detect neuraminidase levels as low as 0.2mU/mL
  • Format allows for multiple time point measurements
  • Designed for minimal autofluorescence interference
  • Detect neuraminidase activity in biological samples such as serum
  • Custom assay design and packaging are also available

Use Amplex Red Assays for a Broad Range of Investigations
A wide variety of validated Amplex Red assays are available for studying cell signaling and lipids, neurobiology, inflammation and immune function, and metabolism. We also offer Amplex UltraRed Reagent, a second-generation reagent providing greater sensitivity and brighter fluorescence, and the Amplex Red/UltraRed Stop Reagent. The Amplex Red/UltraRed Stop Reagent provides convenience and control by allowing the fluorescence signal-generating reaction to be terminated at a user-determined time point. After addition of the stop reagent, the fluorescence signal remains stable for at least three hours.

Enzyme and Protein Activity Assays, Glycosidase, Neuraminidase, and Related Enzyme Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Neuraminidase (Sialidase) Assay
For Use With (Equipment) Microplate Reader, Spectrophotometer, Fluorometer
Product Type Amplex Red Assay Kit
Dye Type Other Label(s) or Dye(s)
Format Tube(s), 96-well plate
Product Line Amplex
Quantity 400 Assays
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.