missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Amplex™ Red Monoamine Oxidase Assay Kit

Product Code. 10114432
Change view
Click to view available options
Quantity:
500 Assays
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10114432 500 Assays Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10114432 Supplier Invitrogen™ Supplier No. A12214

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Ideal for enzyme and protein activity assays

Provides a sensitive and simple fluorometric method for detecting monoamine oxidase activity in purified enzyme preparations at levels as low as 12μU/mL and in tissue samples with as little as 20μg protein in a 100μL assay volume.

Enzyme and Protein Activity Assays, Oxidase and Peroxidase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Monoamine Oxidase Assay
For Use With (Equipment) Fluorometer, Microplate Reader
Product Type Amplex Red Assay Kit
Dye Type Other Label(s) or Dye(s)
Format Cuvettes, 96-well plate
Product Line Amplex
Quantity 500 Assays
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.