missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Vybrant™ Alexa Fluor™ 594 Lipid Raft Labeling Kit

Product Code. 10788463
Click to view available options
Quantity:
1 kit
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10788463

Brand: Invitrogen™ V34405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Provides convenient, reliable and extremely bright fluorescent labeling of lipid rafts in live cells

Vybrant Lipid Raft Labeling Kits include cholera toxin subunit B (CT-B) labeled with either the green-fluorescent Alexa Fluor 488, the orange-fluorescent Alexa Fluor 555 or the red-fluorescent Alexa Fluor 594 dye, as well as an antibody that specifically recognizes CT-B. A 10X solution of phosphate-buffered saline (10X PBS), pH 7.2, and detailed instructions are also included in the kits.

Cell Analysis, Cell Structure, Cell Tracing & Tracking, Lipid Rafts, Organelle Tracing

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Quantity 1 kit
Product Line Alexa Fluor
Protein Subtype Cholera Toxin
Shipping Condition Wet Ice
Label Type Alexa Fluor Dyes
Product Type Lipid Raft Labeling Kit
SubCellular Localization Cell Membranes and Lipids

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.