missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 1,8-ANS (1-Anilinonaphthalene-8-Sulfonic Acid), high purity

Product Code. 11534107
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11534107

Brand: Invitrogen™ A47

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

A sensitive indicator of protein folding, conformational changes and other processes that modify the exposure of the probe to water

  • Essentially nonfluorescent in water, only becoming appreciably fluorescent when bound to membranes

Cell Analysis, Cell Structure, Membranes (General) and Lipids

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Content And Storage Store at room temperature and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Product Type 1,8-ANS
Quantity 100 mg
Shipping Condition Room Temperature
Sub Cellular Localization Cell Membranes & Lipids

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.