Learn More
Emma Safety Footwear Modena Safety Shoes
Water resistant leather uppers, zero penetration in-sole and a steel safety toe cap. Offering marginally more ankle support, the Modena is a shoe that's specifically designed for standing, walking and the occasional run.
Brand:  Emma Safety Footwear MM13609048
Additional Details : Weight : 1.96500kg
 View more versions of this product
Product Code. 15978221
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • Nitrile Rubber Heat Resistant Outsole
  • ESD Certified
  • Chrome-free lining
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011, EN 12568:2010
Black
Hydro-Tec™ Sanitized Silver & Wellness leather
48
Laces
Modena
Leather
Steel