missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Model CHA6 Tychem™ F Hooded Coverall
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 20
Specifications
Specifications
| Product Type | Hooded Coveralls |
| Size | Large |
| Material | Tychem |
| Color | Gray |
| Waist Style | Elastic |
| Ankle Style | Elastic, Attached Socks, Boot Overflaps |
| Cuff Style | Double |
| Neck Style | Hooded |
| Hood Type | Elastic |
| Closure Type | Zipper with Storm Flap |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction