Learn More
DuPont™ Model CHA6 Tychem™ F Hooded Coverall
Product Code. 12653660
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 20
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 12653660
Brand: DuPont™ D13395268
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Specifications
Product Type Hooded Coveralls
Size 2X-Large
Material Tychem
Color Gray
Waist Style Elastic
Ankle Style Elastic, Attached Socks, Boot Overflaps
Cuff Style Double
Neck Style Hooded
Hood Type Elastic
Closure Type Zipper with Storm Flap
Seam Type Stitched, Taped
Protection Class Category III, Type 3-B, 4-B, 5-B and 6-B
Resistance Chemicals
Collar Type Self-adhesive chin flap
Description Tychem™ F Fabric
Size (Metric) Chest 116 to 124 cm
For Use With (Application) For a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications