missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem 2000 C with socks
381.14 GBP valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Use promo code "21673" to get your promotional price.
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Hooded coverall with elastic face opening and adhesive chin flap for tight fit around respirator
- Stitched and over-taped seams
- Double zipper with self-adhesive flap
- Elastic cuffs, ankles and waist,
- Includes thumb loops, integrated socks and boot flaps
- 100% particle tight fabric
- Excellent barrier against a broad range of concentrated inorganic chemicals and biological hazards
- Antistatic treatment (inside)
Specifications
Product Type | Hooded Coveralls |
Size | 2X-Large |
Material | Tychem |
Color | Yellow |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Hood Type | Elastic |
Closure Type | Double Zipper with Self-adhesive Flap |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'