Learn More
Tuttnauer Mobile Lifting Device for 38XX
Product Code. 17809548 Shop All Tuttnauer Products
Change view
Click to view available options
Product Type:
Lifting Device
Unit Size:
Each
Product Code. Product Type unitSize
17809548 Lifting Device Each
1 options
Product Code. 17809548 Supplier Tuttnauer Supplier No. HLD3870026
  • ONLINE EXCLUSIVE PRICE
    £10269.00
    £8405.00 / Pack of 1
    Save £1864.00
    18% Off
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
Mobile lifting device for vertical autoclaves with heavy loads up to 100 kg. The lift is mobile, equipped with an integrated swivel arm for maximum maneuverability and can service a few autoclaves. Controlled by push buttons on a movable handset. Equipped with a maintenance free electrical battery.
  • The mobile lifting device for vertical autoclave basket with heavy sterilization load up to 100 kg.
  • The lift is mobile equipped with an integrated swivel arm for maximum maneuverability and can service a few autoclaves.
  • Controlled by push buttons on a movable control handset.
  • Equipped with a maintenance free electrical battery.
Specifications
Product Type Lifting Device
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.