Learn More
Macherey-Nagel™ MN 96 Bead Plate Type D
Description
- Rack of prefilled tube strips (8 x 12)
- Prefilled with 3 mm steal beads for homogenization
Specifications
Specifications
| Product Type | Rack of Prefilled Tube Strips |
| For Use With (Application) | Sample Homogenization |
| For Use With (Equipment) | Bead Beating |
| Includes | 3 mm steel beads |
| Material | Steel |
| Packaging Quantity | 4 x 96-well Plates |