missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP20 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-62322
This item is not returnable.
View return policy
Description
MMP20 Polyclonal specifically detects MMP20 in Human samples. It is validated for Western Blot.
Specifications
| MMP20 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| AI2A2, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.22, EC 3.4.24.35, Enamel metalloproteinase, Enamelysin, matrix metallopeptidase 20, matrix metalloproteinase 20 (enamelysin), matrix metalloproteinase-20, MMP-20 | |
| Rabbit | |
| 43 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| O60882 | |
| MMP20 | |
| Synthetic peptides corresponding to MMP20(matrix metallopeptidase 20) The peptide sequence was selected from the middle region of MMP20. Peptide sequence AAVYLREPQKTLFFVGDEYYSYDERKRKMEKDYPKNTEEEFSGVNGQIDA. | |
| Affinity purified | |
| RUO | |
| 9313 | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction