missing translation for 'onlineSavingsMsg'
Learn More

MMP Inhibitor II, Thermo Scientific™

Product Code. p-7082466
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15437009 10 mg 10mg
15427009 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437009 Supplier Thermo Scientific Alfa Aesar Supplier No. J65638.MA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A potent, reversible, broad range inhibitor of matrix metalloproteinases

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Description Off-white
Content And Storage -30°C to -10°C
For Use With (Application) A potent, reversible, broad range inhibitor of matrix metalloproteinases
CAS 203915-59-7
Molecular Formula C21H27N3O8S2
Formula Weight 513.58g/mol
Synonym N-Hydroxy-1,3-bis(4-methoxyphenylsulfonyl)-5,5-dimethylhexahydropyrimidine-2-carboxamide
Solubility Soluble in DMSO
Product Type Signal Transduction Reagent
Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.