missing translation for 'onlineSavingsMsg'
Learn More

MMP-9 (92kDa Collagenase IV) Ab-4 Mouse Monoclonal Antibody, Epredia™

Epredia™ MMP-9 (92kDa Collagenase IV) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

£117.30 - £640.00

Specifications

Antigen MMP-9 (92kDa Collagenase IV) Ab-4
Clone IA5
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Quantity Concentration Price Quantity & Availability  
Product Code Brand Quantity Concentration Price Quantity & Availability  
12632387
View Documents
Epredia
MS-816-P0
100 μL 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12642387
View Documents
Epredia
MS-816-P1
500 μL 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12622387
View Documents
Epredia
MS-816-P
1 mL 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12652387
View Documents
Epredia
MS-816-P1ABX
100 μL 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12662387
View Documents
Epredia
MS-816-PABX
200 μL 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

MMPs are a group of enzymes involved in matrix degradation. They share some important characteristics, such as a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and inhibition by specific proteinase inhibitors known as tissue inhibitors of metalloproteinases (TIMPs). Reportedly, MMPs play a crucial role in tumor cell invasion and metastasis.

Host Species: Mouse

Clone: IA5

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human native 92kDa MMP-9

Molecular Weight: 92kDa (proform) and ∽86kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) human fibrosarcoma HT-1080 cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Not suitable)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

MMP-9 (92kDa Collagenase IV) Ab-4
Immunofluorescence
Unconjugated
Mouse
Proteases And Inhibitors
Human native 92kDa MMP-9
Primary
IA5
Monoclonal
Concentrated
RUO
Human
IgG2a κ
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.