missing translation for 'onlineSavingsMsg'
Learn More

MMP-9 (92kDa Collagenase IV) Ab-4 Mouse Monoclonal Antibody, Epredia™

Epredia™ MMP-9 (92kDa Collagenase IV) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-816-P

 View more versions of this product

Product Code. 12622387

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

MMPs are a group of enzymes involved in matrix degradation. They share some important characteristics, such as a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and inhibition by specific proteinase inhibitors known as tissue inhibitors of metalloproteinases (TIMPs). Reportedly, MMPs play a crucial role in tumor cell invasion and metastasis.

Host Species: Mouse

Clone: IA5

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human native 92kDa MMP-9

Molecular Weight: 92kDa (proform) and ∽86kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) human fibrosarcoma HT-1080 cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Not suitable)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

MMP-9 (92kDa Collagenase IV) Ab-4
Monoclonal
200μg/mL
Mouse
1 mL
Proteases And Inhibitors
Human
IgG2a κ
Immunofluorescence
IA5
Unconjugated
Human native 92kDa MMP-9
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.