missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLXIPL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | MLXIPL |
|---|---|
| Concentration | 0.28 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16821384
|
Proteintech
13256-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16811384
|
Proteintech
13256-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ChREBP is a transcription factor involved in activating genes that encode enzymes of fatty acid biosynthesis in liver and adipose. This protein may have relevance to lipid metabolism, obesity, and type II diabetes.Specifications
| MLXIPL | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9NP71 | |
| 51085 | |
| MLXIPL Fusion Protein Ag3481 | |
| Primary | |
| -20°C | |
| MLXIPL |
| 0.28 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| bHLHd14, CHREBP, MIO, MLX interacting protein like, MLX interactor, MLXIPL, MONDOB, WBSCR14, WS bHLH | |
| MLXIPL | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title