missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLXIPL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13256-1-AP-150UL
This item is not returnable.
View return policy
Description
ChREBP is a transcription factor involved in activating genes that encode enzymes of fatty acid biosynthesis in liver and adipose. This protein may have relevance to lipid metabolism, obesity, and type II diabetes.Specifications
| MLXIPL | |
| Polyclonal | |
| Unconjugated | |
| MLXIPL | |
| bHLHd14, CHREBP, MIO, MLX interacting protein like, MLX interactor, MLXIPL, MONDOB, WBSCR14, WS bHLH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51085 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NP71 | |
| MLXIPL | |
| MLXIPL Fusion Protein Ag3481 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction