missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MLKL Recombinant Rabbit Monoclonal Antibody (23GB2935), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | MLKL |
|---|---|
| Clone | 23GB2935 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089071
![]() |
Invitrogen™
MA553518 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088087
|
Invitrogen™
MA553519 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MLKL Recombinant Monoclonal antibody specifically detects MLKL in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| MLKL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q8NB16 | |
| 197259 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2935 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 9130019I15Rik; hMLKL; mixed lineage kinase domain like pseudokinase; mixed lineage kinase domain-like; mixed lineage kinase domain-like protein; Mlkl; rCG51567-like | |
| A synthesized peptide derived from MLKL (1-200AA). | |
| Primary | |
| -20°C | |
| Mlkl |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title