missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MLKL Recombinant Rabbit Monoclonal Antibody (23GB2935), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553518
This item is not returnable.
View return policy
Description
The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.
Specifications
| MLKL | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8NB16 | |
| Mlkl | |
| A synthesized peptide derived from MLKL (1-200AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2935 | |
| Unconjugated | |
| Mlkl | |
| 9130019I15Rik; hMLKL; mixed lineage kinase domain like pseudokinase; mixed lineage kinase domain-like; mixed lineage kinase domain-like protein; Mlkl; rCG51567-like | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 197259 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction