missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML 204
GREENER_CHOICE

Product Code. 30001594
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001594 50 mg 50mg
30001573 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001594 Supplier Tocris Bioscience™ Supplier No. 4732/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective TRPC4 blocker

Blocker of TRPC4 channels (IC50 values are 0.96 and 2.6 μM in fluorescent and electrophysiological assays, respectively). Exhibits 19-fold selectivity against TRPC6 and 9-fold selectivity against TRPC5; displays no significant activity at TRPV1, TRPV3, TRPA1 and TRPM8 channels at concentrations up to 22 μM.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5465-86-1
Molecular Formula C15H18N2

Specifications

Quantity 50 mg
Formula Weight 226.32
Purity >99%
Molecular Weight (g/mol) 226.32
Chemical Name or Material ML 204
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.