CAS RN 5465-86-1
CAS RN 5465-86-1
IUPAC Name:
4-methyl-2-(piperidin-1-yl)quinoline
Synonyms:
4-methyl-2-(piperidin-1-yl)quinoline
Molecular Weight (g/mol):
226.32
Molecular Formula:
C15H18N2
InChi Key:
USYRQXDHKXGTCK-UHFFFAOYSA-N
SMILES:
CC1=C2C=CC=CC2=NC(=C1)N1CCCCC1
1
–
4
of
4
results
1
Tocris Bioscience™ ML 204
CAS: 5465-86-1 Molecular Formula: C15H18N2
2
4-Methyl-2-(1-piperidinyl)-quinoline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Methyl-2-(1-piperidinyl)-quinoline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
ML204, MedChemExpress
MedChemExpress ML204 is a potent, selective TRPC4/TRPC5 channel inhibitor, with at least 19-fold selectivity against TRPC6 and no appreciable effect on all other TRP channels, nor on voltage-gated sodium, potassium, or Ca2+ channels.