missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 133 hydrochloride
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective blocker of inwardly rectifying Kir2 potassium channels (IC50 values are 1.8, 2.8, 2.9 and 4.0 μM at pH 7.4 for mKir2.1, hKir2.6, hKir2.2 and hKir2.3 respectively). Exhibits no effect on rKir1.1 (IC50 > 300 μM); displays weak activity at hKir7.1 and rKir4.1 (IC50 values are 33 and 76 μM).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 313.82 |
| Purity | >99% |
| Molecular Weight (g/mol) | 313.83 |
| Chemical Name or Material | ML 133 hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction