CAS RN 1222781-70-5
IUPAC Name:
[(4-methoxyphenyl)methyl][(naphthalen-1-yl)methyl]amine hydrochloride
Synonyms:
[(4-methoxyphenyl)methyl](naphthalen-1-ylmethyl)amine hydrochloride
Molecular Weight (g/mol):
313.83
Molecular Formula:
C19H20ClNO
InChi Key:
NGQIBUUFXDPHKT-UHFFFAOYSA-N
SMILES:
Cl.COC1=CC=C(CNCC2=C3C=CC=CC3=CC=C2)C=C1
1
–
4
of
4
results
1
Tocris Bioscience™ ML 133 hydrochloride
CAS: 1222781-70-5 Molecular Formula: C19H20ClNO
2
ML 133 Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
ML 133 Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
ML133 hydrochloride, MedChemExpress
MedChemExpress ML133 hydrochloride is a selective Kir2 family channels inhibitor, with an IC50 of 1.8 μM at pH 7.4 and 290 nM at pH 8.5.