missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKS1 Mouse anti-Human, Porcine, Rat, Clone: 6A8B11, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | MKS1 |
|---|---|
| Clone | 6A8B11 |
| Concentration | 0.9 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16888465
|
Proteintech
66518-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16878465
|
Proteintech
66518-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene localizes to the basal body and is required for formation of the primary cilium in ciliated epithelial cells. Mutations in this gene result in Meckel syndrome type 1 and in Bardet-Biedl syndrome type 13. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| MKS1 | |
| 0.9 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| BBS13, Meckel syndrome type 1 protein, Meckel syndrome, type 1, MES, MKS, MKS1 | |
| Mks1 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 6A8B11 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Rat, Pig | |
| Q499Q5, Q9NXB0 | |
| 100514538, 287612, 54903 | |
| MKS1 Fusion Protein Ag9504 | |
| Primary | |
| -20°C | |
| MKS1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title