missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKS1 Mouse anti-Human, Porcine, Rat, Clone: 6A8B11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66518-1-IG-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene localizes to the basal body and is required for formation of the primary cilium in ciliated epithelial cells. Mutations in this gene result in Meckel syndrome type 1 and in Bardet-Biedl syndrome type 13. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| MKS1 | |
| Monoclonal | |
| 0.9 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q499Q5, Q9NXB0 | |
| Mks1 | |
| MKS1 Fusion Protein Ag9504 | |
| 20 μL | |
| Primary | |
| Human, Rat, Pig | |
| Antibody | |
| IgG1 |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 6A8B11 | |
| Unconjugated | |
| MKS1 | |
| BBS13, Meckel syndrome type 1 protein, Meckel syndrome, type 1, MES, MKS, MKS1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100514538, 287612, 54903 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction