missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKP-2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1G1C12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66349-1-IG-150UL
This item is not returnable.
View return policy
Description
DUSP4 is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP) kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which are associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. DUSP4 inactivates ERK1, ERK2 and JNK, is expressed in a variety of tissues, and is localized in the nucleus.Specifications
| MKP-2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13115, Q62767, Q8BFV3 | |
| Dusp4 | |
| MKP-2 Fusion Protein Ag1206 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1G1C12 | |
| Unconjugated | |
| DUSP4 | |
| dual specificity phosphatase 4, DUSP4, HVH2, MAP kinase phosphatase 2, MKP 2, MKP2, MKP-2, TYP, VH2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100514374, 1846, 319520, 60587 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction